missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Valine, +99%

Product Code. 10183850
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-06-3
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00004267
InChI Key KZSNJWFQEVHDMF-UHFFFAOYSA-N
Synonym dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid
PubChem CID 1182
ChEBI CHEBI:27266
IUPAC Name 2-amino-3-methylbutanoic acid
SMILES CC(C)C(N)C(O)=O

Specifications

Melting Point 283.5°C to 285.0°C
Color White
Chlorine (Cl) 400ppm max.
Heavy Metals (as Pb) 10ppm max.
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula (CH3)2CHCH(NH2)CO2H
Beilstein 04, 430
Loss on Drying 0.2% max. (105°C, 3 hrs)
Merck Index 14, 9909
Packaging Plastic Bottle
Solubility Information Solubility in water: 68g/L. Other solubilities: practically insoluble in cold alcohol and ether, soluble in mineral acids, insoluble in organic solvents, slightly soluble in benzene, ethanol
Formula Weight 117.15
Transmittance 95% min. (c=10, 2N HCl, 430nm) 1cm cell
Percent Purity ≥99%
Sulfated Ash 0.2% max.
Grade Biochemical
Density 1.3100g/mL
Quantity 250 g
Specific Gravity 1.31
Physical Form Crystalline Powder
Chemical Name or Material DL-Valine, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'