missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Valine, Thermo Scientific™
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
Specifications
Specifications
Packaging | Plastic Bottle |
Formula Weight | 117.15 |
Grade | Extra Pure |
Quantity | 1 kg |
Chemical Name or Material | DL-Valine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'