Learn More
DLin-KC2-DMA, MedChemExpress
MedChemExpress DLin-KC2-DMA is a cationic/ionizable lipid for siRNA delivery.
Brand:  Medchem Express HY-112758
Encompass_Preferred
 View more versions of this product
Product Code. 16333237
  • POA / 100mg
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
DLin-KC2-DMA is a cationic/ionizable lipid for siRNA delivery.
Specifications
Specifications
DLin-KC2-DMA
Colorless
C43H79NO2
Ethanol : 100 mg/mL (155.74 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (155.74 mM; Need ultrasonic)
642.09
98.0%
Liquid
1190197-97-7
Research
100 mg
CCCCC/C=C\C/C=C\CCCCCCCCC1(CCCCCCCC/C=C\C/C=C\CCCCC)OC(CCN(C)C)CO1
642.09
Research
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
DLin-KC2-DMA, MedChemExpress