Learn More
Leica Microsystems DM IL LED Microscope
Ergonomically designed inverted microscope with high-performance optics. Leica Microsystems™ DM IL LED Microscope is ideal for cell culture examination and micromanipulation.
Brand:  Leica Microsystems DM-IL-LEDTM
 View more versions of this product
Product Code. 12868035
  • £8180.00 / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for applications in cell biology and medicine, micromanipulation, biotechnology, developmental biology, transgenics, molecular biology and fluorescence.
  • Ergonomic arrangement of controls for fatigue free operation.
  • In addition to phase contrast and brightfield illumination, the DM IL features Leica's unique integrated modulation contrast (IMC) without the need for special objectives.
    • Available for 10x, 20x, 32x, and 40x magnification.
  • Comprehensive set of contrast methods.
  • LED illumination provides low energy consumption and no heat build up.
  • LEDs last 50,000 hours.
  • Fluorescence capable (LF Model)
  • Sturdy design with low stage height and plenty of operation space.
  • Long working distances to accommodate large culture flasks.
  • Can be upgraded to accommodate large-scale cell production.
  • Certified for in-vitro diagnostics (IVD) including in-vitro fertilization (IVF).
Specifications
Specifications
No fluorescence accessories.
Brightfield Microscopy
Inverted Microscope
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products