missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMAC, Dimethylacetamide, GC Headspace Grade, Thermo Scientific™
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Description
- Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
- 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
- This product is UV Absorbance Tested to ensure low organic contamination
- Processed for low water content to facilitate the extraction of organic volatile impurities, or VOC

Specifications
Specifications
Absorbance | 0.300 AU Maximum (275nm), 0.080 AU Maximum (300nm), 0.005 AU Maximum (350nm) |
CAS | 127-19-5 |
Color | Colorless |
Quantity | 1 L |
Identification | Pass Test |
Packaging | Glass Bottle |
Physical Form | Liquid |
Molecular Formula | C4H9NO |
Titratable Acid | 0.001 mEq/g Maximum |
MDL Number | MFCD00008686 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
245156 | Certificate of Analysis | 31/01/2025 | D160-1 |
Safety and Handling


Product Identifier
- N,N-Dimethylacetamide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Flammable liquid Category 3
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
- H373-May cause damage to organs.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : dimethyl acetamide
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
6Minutes
33Seconds
57
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
DMAC, Dimethylacetamide, GC Headspace Grade, Thermo Scientific™ >
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu