missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ DMEM/F-12, HEPES

Click to view available options
Quantity:
10 x 500 mL
500 mL
Unit Size:
5000mL
500mL
Description
Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM / F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM / F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of Gibco™ DMEM / F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool. Gibco™ DMEM / F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DMEM / F-12 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ DMEM / F-12 product made in our Grand Island facility (11330-057). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
DMEM / F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM / F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM / F-12 uses a sodium bicarbonate buffer system and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DMEM / F-12 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ DMEM / F-12 product made in our Grand Island facility (11330-057). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
DMEM / F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM / F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM / F-12 uses a sodium bicarbonate buffer system and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
Specifications
| Concentration | 1 X |
| Form | Liquid |
| Product Type | DMEM/F-12 Medium |
| With Additives | Glutamine, HEPES, Phenol Red, Sodium Pyruvate |
| For Use With (Application) | Mammalian cell culture |
| Quantity | 10 x 500 mL |
| Certifications/Compliance | ISO 13485 standards, FDA |
| pH | 6.9 to 7.3 |
| Regulatory Status | For in-vitro diagnostic use |
| Content And Storage | 2 to 8°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction