missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DMEM/F-12, no glutamine
GREENER_CHOICE

Product Code. 11530566
Click to view available options
Quantity:
500 mL
10 x 500 mL
Unit Size:
5000mL
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11530566

Brand: Gibco™ 21331020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM/F-12) is a widely used basal medium for supporting the growth of many different mammalian cells.

Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM/F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of Gibco™ DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM/F-12 is modified as follows:

With: Phenol Red

Without: L-glutamine, HEPES

Gibco™ DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids and vitamins with F-12's wide variety of components.

DMEM/F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM/F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5-10% CO2 environment to maintain physiological pH.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mammalian Cells
Concentration 1 X
Form Liquid
Product Type Dulbecco's Modified Eagle Medium (DMEM)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
With Additives High Glucose, Phenol Red, Sodium Pyruvate
Without Additives No Glutamine, No HEPES
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.