Learn More
Gibco™ DMEM/F-12, no glutamine
Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM/F-12) is a widely used basal medium for supporting the growth of many different mammalian cells.
Brand: Gibco™ 21331046
118.28 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.52500kg
Description
Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM/F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of Gibco™ DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This DMEM/F-12 is modified as follows:
With: Phenol Red
Without: L-glutamine, HEPES
Gibco™ DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids and vitamins with F-12's wide variety of components.
DMEM/F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM/F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Especificaciones
ISO 13485 standards, FDA | |
DMEM/F-12 Medium, 500mL, Liquid Form, With Phenol Red, Sodium Pyruvate, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture | |
DMEM (Dulbecco's Modified Eagle Medium)/F-12 | |
No Glutamine, No HEPES | |
cGMP-compliant under the ISO 13485 standard | |
For in-vitro diagnostic use |
2 to 8°C | |
Liquid | |
Phenol Red, Sodium Pyruvate | |
Mammalian cell culture | |
10 x 500 mL |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.