Learn More
Gibco™ DMEM/F-12, no phenol red
GREENER_CHOICE
DMEM/F-12, no phenol red
Brand:  Gibco™ 21041033
Product Code. 12088089
  • £195.00 / 5000mL
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
Using DMEM/F-12
DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids and vitamins with F-12's wide variety of components. DMEM/F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM/F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5–10% CO2 environment to maintain physiological pH.
cGMP manufacturing and quality system
For supply chain continuity, we manufacture DMEM/F-12 at two separate facilities, located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP-manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 13485 standards, FDA
DMEM Medium, 500mL, 1 X Concentration, Liquid Form, With L-glutamine, Sodium Pyruvate, 7 pH, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Soluble in Water, Used for Mammalian Cell Culture
Liquid
Glutamine, Sodium Pyruvate
Mammalian cell culture
10 x 500 mL
2 to 8°C
1 X
DMEM Medium
No HEPES, No Phenol Red
7