Learn More
Gibco™ DMEM/F-12, powder
DMEM/F-12, powder
Brand:  Gibco™ 32500043
 View more versions of this product
Product Code. 11504466
  • £116.62 / 50 L
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Dulbecco's Modified Eagle Medium:Nutrient Mixture F-12 (DMEM / F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM / F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of Gibco™ DMEM / F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool. Gibco™ DMEM / F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DMEM / F-12 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ DMEM / F-12 product made in our Grand Island facility (12500-096). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
DMEM / F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM / F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM / F-12 uses a sodium bicarbonate buffer system and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 13485 standards, FDA
DMEM/F-12, 50L, Powder, 6.2 to 6.6 pH, 250 to 290mOsm/kg Osmolality, 36 Months Shelf Life, Ambient Shipping Condition, 2 to 8°C Storage Requirements, Animal Origin-Free Classification, For Mammalian Cell Culture
DMEM/F-12 Medium
No HEPES, No Sodium Bicarbonate
6.2 to 6.6
CE-IVD
2 to 8°C, room temperature
Powder
Glutamine, Phenol Red, Sodium Pyruvate
Mammalian cell culture
50 L