missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DMEM, high glucose, GlutaMAX™ Supplement, pyruvate
GREENER_CHOICE

DMEM, high glucose, GlutaMAX™ Supplement, pyruvate

£17.53 - £154.00

Specifications

Certifications/Compliance ISO 13485 standards, FDA
Content And Storage 2 to 8°C
Concentration 1 X
Form Liquid
Product Type Dulbecco's Modified Eagle Medium
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11594446
GREENER_CHOICE
View Documents Promotion Details
Gibco™
31966021
500 mL
£17.53
500mL
Please to purchase this item. Need a web account? Register with us today!
12077549
GREENER_CHOICE
View Documents
Gibco™
31966047
10 x 500 mL
£154.00
5000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

Using DMEM
DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Product use
For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal law restricts this device to sale by or on the order of a physician. Customers using Gibco™ DMEM in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP manufacturing and quality system
DMEM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical DMEM product made in our Scotland facility (31966-021). This facility is also registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standard.
Specifications

Specifications

ISO 13485 standards, FDA
1 X
Dulbecco's Modified Eagle Medium
No HEPES
6.8 to 7.2
2 to 8°C
Liquid
High Glucose, Sodium Pyruvate, GlutaMAX, Phenol Red
Tissue and cell culture processing, mammalian cell culture
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.