missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DMEM, low glucose, GlutaMAX™ Supplement, pyruvate
GREENER_CHOICE

Product Code. p-7075810
Click to view available options
Quantity:
10 x 500 mL
500 mL
Unit Size:
5000mL
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11580586

Brand: Gibco™ 21885108

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

DMEM, low glucose, GlutaMAX™ Supplement, pyruvate

Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. Gibco™ DMEM with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ DMEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ DMEM product made in our Grand Island facility (10567-022). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1X
Form Liquid
Product Type DMEM (Dulbecco's Modified Eagle Medium)
With Additives Low Glucose, Sodium Pyruvate, GlutaMAX, Phenol Red
Without Additives No HEPES
For Use With (Application) Mammalian cell culture
Quantity 10 x 500 mL
Certifications/Compliance ISO 13485 standards, FDA
pH 7 to 7.2
Regulatory Status For in-vitro diagnostic use
Content And Storage Store at 2–8°C.
Description DMEM Medium, 500mL, 1 X Concentration, Liquid Form, With Low Glucose, Sodium Pyruvate, GlutaMAX™, Phenol Red, 7 to 7.2 pH, 12 Months Shelf Life, 300 to 340mOsm/kg Osmolality, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.