Learn More
Gibco™ DMEM, low glucose, pyruvate
GREENER_CHOICE
DMEM, low glucose, pyruvate
Brand:  Gibco™ 31885049
 View more versions of this product
Product Code. 11574446
  • £138.00 / 5000mL
Edge
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DMEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ DMEM product made in our Grand Island facility (11885-092). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 13485 standards, FDA
Dulbecco's Modified Eagle Medium (DMEM), 10 x 500mL, Liquid, 1 X Concentration, 7.0 to 7.4 pH, With Low Glucose, L-Glutamine, Sodium Pyruvate, Phenol Red, 12 Months Shelf Life, 300 to 340mOsm/kg Osmolality, For Mammalian Cell Culture
Liquid
Low Glucose, Glutamine, Sodium Pyruvate, Phenol Red
Mammalian cell culture
10 x 500 mL
2 to 8°C
1 X
Dulbecco's Modified Eagle Medium
No HEPES
7.0 to 7.4