missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ DMEM, low glucose, pyruvate, no glutamine, no phenol red
DMEM, low glucose, pyruvate, no glutamine, no phenol red
Brand: Gibco™ 11880036
109.25 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
This item is not returnable.
View return policy
Description
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
cGMP Manufacturing and Quality System
Gibco™ DMEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DMEM product made in our Grand Island facility (11054-020). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
DMEM Medium, 500mL, 1 X Concentration, Liquid Form, With Low Glucose, Sodium Pyruvate, 7 to 7.4 pH, 12 Months Shelf Life, 300 to 340mOsm/kg Osmolality, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture | |
Liquid | |
Low Glucose, Sodium Pyruvate | |
Mammalian cell culture | |
10 x 500 mL |
2 to 8°C | |
1 X | |
DMEM Medium | |
No Phenol Red, No Glutamine, No HEPES | |
7 to 7.4 | |
For research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction