missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMF, Dimethylformamide, Sequencing, Fisher BioReagents
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
Description
- DMF for sequencing and peptide synthesis
- Useful for protein chemistry due to low amine content
- DMF is a commonly used solvent in solid phase peptide synthesis
- Intended for research use only
Specifications
Specifications
pH | 6 to 8 |
Melting Point | -61°C |
Absorbance | Pass Test, 1 max. at 270nm, 0.3 max. at 275nm, 0.1 max. at 295nm, 0.05 max. at 310nm, 0.01 max. (340nm to 400nm) at 340nm |
Color | Colorless |
Decomposition Information | >350°C |
Density | 0.945g/cm³ |
Boiling Point | 153°C |
Quantity | 500 mL |
Identification | Pass Test |
Packaging | Glass Bottle |
Show More |
Safety and Handling
Product Identifier
- N,N-Dimethylformamide
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'