Learn More
Tocris Bioscience™ DMNB-caged-Serine
Caged serine; excited by visible blue light
Brand: Tocris Bioscience™ 6315/50
Additional Details : Weight : 0.00970kg
Description
DMNB-caged-Serine is a DMNB-caged serine. Photoactivated to release serine by visible blue light (405 nm) in cells. Incorporated into S.cerevisiae proteins when introduced into growth medium at millimolar concentrations.Specifications
O-(4,5-Dimethoxy-2-nitrobenzyl)-L-serine | |
50 mg | |
C12H16N2O7 |
780009-55-4 | |
Caged Compounds | |
98% |