missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMSO, sterile filtered, Tocris Bioscience™
Click to view available options
Quantity:
25 mL
100 mL
Description
DMSO, sterile filtered is a solvent with wide ranging applications in cell culture, cryopreservation and organic chemistry. DMSO dissolves both polar and nonpolar compounds. Improves responsiveness of human embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) to differentiation signals. DMSO is used in PCR to inhibit secondary structure formation by DNA template or primers.
Specifications
Quantity | 25 mL |
Formula Weight | 78.13 |
Percent Purity | >99.9% |
Chemical Name or Material | DMSO, sterile filtered |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'