Learn More
Thermo Scientific™ DNA AWAY™ Surface Decontaminant
Eliminate unwanted DNA and DNase from glassware and plasticware without affecting subsequent DNA samples. This surface decontaminant degrades DNA more quickly and effectively than autoclaving.
Brand: Thermo Scientific™ 7010
16.95 GBP valid until 2025-06-30
Use promo code "25076" to get your promotional price.
Description
Eliminate unwanted DNA and DNase from glassware and plastic ware without affecting subsequent DNA samples. Thermo Scientific™ DNA AWAY™ Surface Decontaminant degrades DNA more quickly and effectively than autoclaving.
- Suitable for gel boxes, pipettes, benchtops, thermal cyclers, or other apparatus
- Use before performing PCR or working with DNA for a DNA-free work area
- Simply apply the ready-to-use formula to the surface to decontaminate, then wipe dry or rinse clean

Specifications
11 | |
DNA Surface Decontaminant | |
250 mL |
Sodium Hydroxide with Water | |
8.5 oz. |
Safety and Handling
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide
Special Offers
- Multiple Manufacturers
-
Thermo Scientific ClipTip
,
Thermo Scientific ART
,
Thermo Scientific Finnpipette
,
Thermo Scientific Matrix
Exclusive Promotion on Liquid Handling
Unlock Precision: Top Pipettes and Tips on Promotion!Promo Code: 25076Expires: 2025-06-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.