Learn More
MP Biomedicals™ DNA-OFF™
DNA-OFF™ is a ready to use non-alkaline cleaning solution that is highly active against DNA contamination. It is non-carcinogenic, non corosive and heat resistant solution used for eliminating DNA from surfaces, such as PCR workstations, countertops, apparatus and pipettors.
Brand:  MP Biomedicals™ 11QD0500
95.37 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Product Code. 11477130
  • £141.00 / 500mL
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The presence of contaminant DNA at a PCR work area can result in DNA amplification artifacts.
  • This ready-to-use solution is highly active against DNA contamination and is intended for use at PCR workstations, including countertops, apparatus and pipettors.
  • It is stable, heat resistant, non-corrosive and free of carcinogenic compounds.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Removal Agent
500 mL
Intended for use at PCR workstations, including countertops, apparatus and pipettors
16.9 oz.
Highly active non-alkaline coleaning solution for DNA contamination, ready to use.
Bottle