missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DNA-PK Recombinant Protein
DNA-PK is a heterotrimeric, DNA-dependent protein kinase that initiates signalling cascades in response to DNA damage.
£434.00 - £12070.00
Specifications
Gene Alias | DNA-PK |
---|---|
KinaseFamily | PIKK Kinase Family |
Species | Human |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15117975
|
Invitrogen™
PR9107A |
100μg |
£2595.00
100µg |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15378517
|
Invitrogen™
PV5865 |
1mg |
£12070.00
Each |
Estimated Shipment: 22-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15428803
|
Invitrogen™
PV5866 |
10 μg |
£434.00
10µg |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
DNA-PK is an atypical kinase, closely related to the PI3 kinases, and is often potently inhibited by small molecules designed to target PI3 kinases.
Specifications
DNA-PK | |
PIKK Kinase Family | |
Human | |
Baculovirus |