missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA Polymerase I, Large (Klenow) Fragment

DNA Polymerase I, Large (Klenow) Fragment

Brand:  Invitrogen™ 18012096

Product Code. 10023042

  • £730.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

DNA Polymerase I, Large (Klenow) Fragment is a DNA polymerase enzyme that lacks the 5' to 3' exonuclease activity of intact DNA Polymerase I, but does exhibit the 5' to 3' DNA polymerase and 3' to 5' exonuclease activities.

Applications:
Fill-in of 5´ overhangs (1). Synthesis of probes by random primers labeling method (2). Sequencing single- and double-stranded DNA (3). Site-directed mutagenesis.

Source:
Purified from E. coli expressing the Klenow fragment on a plasmid.

Performance and Quality Testing:
SDS-PAGE purity; single-stranded and double-stranded endodeoxyribonuclease and self-priming assays; performance evaluated in fill-in reaction.

Unit Definition:
One unit incorporates 10 nmol of total deoxyribonucleotide into acid-precipitable material in 30 min. at 37°C using a template
  • primer.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    –20°C
    2 x 500 Unit(s)
    DNA polymerase I, large (Klenow) fragment is supplied with a vial of 10X REact™ 2 buffer [500mM Tris HCl (pH 8.0), 100mM MgCl2, 500mM NaCl], vial of dilution buffer
    DNA Polymerase I Large (Klenow) Fragment
    Genotyping, Cloning
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.