missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ DNA Polymerase I (10 U/μL)
DNA Polymerase I is a template-dependent DNA polymerase which catalyzes 5'→3' synthesis of DNA.
£59.50 - £246.00
Specifications
Description | DNA Polymerase I |
---|---|
Polymerase | DNA Polymerase I |
Concentration | 10 U/μL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10364630
|
Thermo Scientific™
EP0041 |
500 U |
£59.50
500 units |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10889010
|
Thermo Scientific™
EP0042 |
2500 U |
£246.00
2500 units |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
DNA Polymerase I, a template-dependent DNA polymerase, catalyzes 5'→3' synthesis of DNA. The enzyme also exhibits 3'→5' exonuclease (proofreading) activity, 5'→3' exonuclease activity, and ribonuclease H activity.
Highlights
- Incorporates modified nucleotides (e.g. biotin-, digoxigenin-, aminoallyl-, fluorescently-labeled nucleotides)
- Active in multiple buffers, including restriction enzyme, PCR, and RT buffers
Specifications
DNA Polymerase I | |
DNA Polymerase I | |
10 U/μL |
For Research Use Only. Not for use in diagnostic procedures.