Learn More
Fisherbrand™ DNA Sequencing Systems: Combs for Discontinued Model FB-SEQ-2045
DNA Sequencing Systems Combs for discontinued model FB-SEQ-2045
£36.40 - £64.35
Product Code | Brand | Thickness (Metric) Teeth | Volume (Metric) Well | Product Type | Teeth Count | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thickness (Metric) Teeth | Volume (Metric) Well | Product Type | Teeth Count | Price | Quantity & Availability | ||
11907505
|
Fisherbrand™
S1S-SHARK2 |
0.2 mm | 5μL | Sharktooth Comb | 39 |
£36.40
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12474707
|
Fisherbrand™
S1S-MT2 |
0.4 mm | 10μL | Sharktooth Micro-Well Comb | 36 |
£64.35
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11933075
|
Fisherbrand™
S1S-20A |
0.4 mm | 15μL | Sharktooth Comb | 20 |
£49.90
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12404717
|
Fisherbrand™
S1S-MT44 |
0.4 mm | 3.7μL | Sharktooth Micro-Well Comb | 73 |
£44.40
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12484707
|
Fisherbrand™
S1S-MT3 |
0.4 mm | 5.5μL | Sharktooth Micro-Well Comb | 55 |
£44.40
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11917505
|
Fisherbrand™
S1S-SHARK4 |
0.4 mm | 9μL | Sharktooth Comb | 39 |
£36.40
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12494707
|
Fisherbrand™
S1S-MT42 |
0.2 mm | 2μL | Sharktooth Micro-Well Comb | 73 |
£44.40
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
For use with discontinued model FB-SEQ-2045
Specifications
Owl s4s aluminum backed sequencer system |