missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£122.57 - £303.85
Specifications
Antigen | DNAJC16 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16857534
|
Proteintech
17599-1-AP-20UL |
20 μL |
£122.57
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16847534
|
Proteintech
17599-1-AP-150UL |
150 μL |
£303.85
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
DNAJC16 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q5FVM7, Q80TN4, Q9Y2G8 | |
214063, 23341, 362652 | |
DNAJC16 Fusion Protein Ag11637 | |
Primary | |
-20°C | |
DNAJC16 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DNAJC16, KIAA0962 | |
Dnajc16 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title