Learn More
Thermo Scientific™ DNAPac™ RP Columns

Get high resolution LC and LC-MS separation of oligonucleotides, including double-stranded (ds) DNA/RNA fragments, using these DNAPac reversed phase columns.
Brand: Thermo Scientific™ 088923
737.42 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Achieve superior reversed phase oligonucleotide separations using the Thermo Scientific™ DNAPac™ RP HPLC columns. The unique chemistry is designed for analysis of oligonucleotides and double-stranded (ds) DNA/RNA fragments using LC-UV or LC-MS. The column chemistry provides excellent performance under a broad range of pH, temperature, and mobile phase compositions. In addition, the wide pore size of the resin provides excellent separation of large double-stranded nucleic acids up to 10k base pairs.
High Performance Reversed-Phase LC ColumnsThe DNAPac RP column is based on spherical wide-pore 4μm polymer resin. The phase is highly stable to extremes of pH (0-14), and temperature (5-110°C), and operates with mobile phases compatible with direct mass spectrometry. The column may be operated in both HPLC and UHPLC instruments, producing high resolution at high throughput for oligonucleotides and resolves both small and very large dsDNA segments. DNAPac RP reversed phase (RP) column is specifically designed for separation of oligonucleotides and DNA/RNA fragments. The stationary phase is fully compatible with mass spectrometry friendly organic solvents and ion-pair reagents such as triethylamine (TEA) and hexylamine (HA). The wide pore size of polymer resin enables efficient separation of large nucleic acids with low carry-over.
Columns designed for reversed phase oligonucleotide LC and LC/MS separations
- Designed for ion-pair reversed-phase (IP-RP) separations of both oligonucleotides and double-stranded nucleic acids
- High resolution and high throughput
- Excellent MS compatibility
- Wide operating pH range (0-14) and high temperature stability (up to 110°C)

Specifications
Analytical Column | |
100 mm | |
4 μm | |
Proprietary Wide Pore | |
<110°C | |
DNAPac | |
Reversed Phase | |
Bio-LC |
2.1 mm | |
4000 psi (275 bar) | |
0 to 14 | |
1 Ea. | |
DVB (Divinylbenzene) Polymer | |
DVB (Divinylbenzene) Polymer | |
Reversed Phase | |
Responsibly packaged |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
28Hours
3Minutes
38Seconds
20 -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
59Hours
3Minutes
38Seconds
20New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.