Learn More
Thermo Scientific™ DNAPac™ RP Columns
Get high resolution LC and LC-MS separation of oligonucleotides, including double-stranded (ds) DNA/RNA fragments, using these DNAPac reversed phase columns.
Brand: Thermo Scientific™ 080922-1521232
6506.39 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Achieve superior reversed phase oligonucleotide separations using the Thermo Scientific™ DNAPac™ RP HPLC columns. The unique chemistry is designed for analysis of oligonucleotides and double-stranded (ds) DNA/RNA fragments using LC-UV or LC-MS. The column chemistry provides excellent performance under a broad range of pH, temperature, and mobile phase compositions. In addition, the wide pore size of the resin provides excellent separation of large double-stranded nucleic acids up to 10k base pairs.
DNAPac RP reversed phase (RP) column is based on spherical wide-pore 4 μm polymer resin specifically designed for separation of oligonucleotides and DNA/RNA fragments. The stationary phase is highly stable to extreme pH (0-14), and temperature (5-110°C) and fully compatible with mass spectrometry friendly organic solvents and ion-pair reagents such as triethylamine (TEA) and hexylamine (HA). The wide pore size of polymer resin enables efficient separation of small to large nucleic acids with low carry-over. The column may be operated in both HPLC and UHPLC instruments, producing high resolution at high throughput for oligonucleotides and resolves both small and very large dsDNA segments:
- Designed for ion-pair reversed-phase (IP-RP) LC and LC/MS separations of both oligonucleotides and double-stranded nucleic acids
- High resolution and high throughput
- Excellent MS compatibility
- Wide operating pH range (0-14) and high temperature stability (up to 110°C)
- Biocompatible hardware to eliminate analyte absorption

Specifications
Preparative column, standard | |
4 to 8 mL/min | |
2900 psi (200 bar) | |
0 to 14 | |
up to 110°C | |
DNAPac | |
Reversed Phase | |
DVB (Divinylbenzene) Polymer | |
Reversed Phase HPLC |
21.2 mm | |
150 mm | |
4 μm | |
Each | |
Proprietary | |
4 µm polymer Resin | |
Reversed Phase | |
Preparative Column |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.