missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNAPac™ RP Columns

Get high resolution LC and LC-MS separation of oligonucleotides, including double-stranded (ds) DNA/RNA fragments, using these DNAPac reversed phase columns.

Brand:  Thermo Scientific™ 080922-0121232

780.94 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 18088201

  • £788.93 / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Achieve superior reversed phase oligonucleotide separations using the Thermo Scientific™ DNAPac™ RP HPLC columns. The unique chemistry is designed for analysis of oligonucleotides and double-stranded (ds) DNA/RNA fragments using LC-UV or LC-MS. The column chemistry provides excellent performance under a broad range of pH, temperature, and mobile phase compositions. In addition, the wide pore size of the resin provides excellent separation of large double-stranded nucleic acids up to 10k base pairs.

DNAPac RP reversed phase (RP) column is based on spherical wide-pore 4 μm polymer resin specifically designed for separation of oligonucleotides and DNA/RNA fragments. The stationary phase is highly stable to extreme pH (0-14), and temperature (5-110°C) and fully compatible with mass spectrometry friendly organic solvents and ion-pair reagents such as triethylamine (TEA) and hexylamine (HA). The wide pore size of polymer resin enables efficient separation of small to large nucleic acids with low carry-over. The column may be operated in both HPLC and UHPLC instruments, producing high resolution at high throughput for oligonucleotides and resolves both small and very large dsDNA segments:

  • Designed for ion-pair reversed-phase (IP-RP) LC and LC/MS separations of both oligonucleotides and double-stranded nucleic acids
  • High resolution and high throughput
  • Excellent MS compatibility
  • Wide operating pH range (0-14) and high temperature stability (up to 110°C)
  • Biocompatible hardware to eliminate analyte absorption
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Semi-preparative Guard Cartridge
4 to 8 mL/min
4000 psi (275 bar)
2 PK.
DNAPac
Reversed Phase
DVB (Divinylbenzene) Polymer
Reversed Phase HPLC
21.2 mm
10 mm
0 to 14
Proprietary
4 µm polymer Resin
Reversed Phase
Semi-preparative
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ DNAPac™ RP Columns >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.