Learn More
Thermo Scientific™ DNase I, RNase-free, HC (50 U/μL)
Endonuclease that nonspecifically cleaves single- and double-stranded DNA to release di-, tri-, and oligonucleotides with 5'-phosphate and 3'-OH groups.
Brand: Thermo Scientific™ EN0523
52.44 GBP valid until 2024-12-31
Use promo code "21884" to get your promotional price.
Code : 88
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific DNase I, RNase-free is an endonuclease that digests single- and double-stranded DNA. It hydrolyzes phosphodiester bonds producing mono- and oligodeoxyribonucleotides with 5'-phosphate and 3'-OH groups.
The enzyme activity is strictly dependent on Ca2+ and is activated by Mg2+ or Mn2+ions.
- In the presence of Mg2+, DNase I cleaves each strand of dsDNA independently in a statistically random fashion
- In the presence of Mn2+, the enzyme cleaves both DNA strands at approximately the same site, producing DNA fragments with blunt-ends or with overhang termini of only one or two nucleotides
Highlights
- Recombinant enzyme
- Purified from non-animal host with a lower level of intrinsic RNases
Applications
- Preparation of DNA-free RNA
- Removal of template DNA followingin vitrotranscription
- Preparation of DNA-free RNA prior to RT-PCR and RT-qPCR
- DNA labeling by nick-translation in conjunction with DNA Polymerase I
- Studies of DNA-protein interactions by DNase I, RNase-free footprinting
- Generation of a library of randomly overlapping DNA inserts. Reaction buffer containing Mn2+ is used
Note
DNase I is sensitive to physical denaturation. Mix gently by inverting the tube. Do not vortex.
Specifications
50 U/μL | |
Reaction Buffer | |
DNase I RNase Free |
DNase | |
1000 U |
For Research Use Only. Not for use in diagnostic procedures.