missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Docosanoic acid, 85%, technical, Thermo Scientific Chemicals
£42.90 - £147.00
Chemical Identifiers
CAS | 112-85-6 |
---|---|
Molecular Formula | C22H44O2 |
Molecular Weight (g/mol) | 340.58 |
MDL Number | MFCD00002807 |
InChI Key | UKMSUNONTOPOIO-UHFFFAOYSA-N |
Synonym | behenic acid, 1-docosanoic acid, n-docosanoic acid, hydrofol acid 560, docosoic acid, hydrofol 2022-55, glycon b-70, behensaeure, docosanoicacid, docosansaeure |
PubChem CID | 8215 |
ChEBI | CHEBI:28941 |
IUPAC Name | docosanoic acid |
SMILES | CCCCCCCCCCCCCCCCCCCCCC(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10676174
|
Thermo Scientific Acros
334812500 |
250 g | Glass bottle |
£42.90
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10451810
|
Thermo Scientific Acros
334810010 |
1 kg | Plastic bottle |
£147.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-85-6 | |
340.58 | |
UKMSUNONTOPOIO-UHFFFAOYSA-N | |
8215 | |
docosanoic acid |
C22H44O2 | |
MFCD00002807 | |
behenic acid, 1-docosanoic acid, n-docosanoic acid, hydrofol acid 560, docosoic acid, hydrofol 2022-55, glycon b-70, behensaeure, docosanoicacid, docosansaeure | |
CHEBI:28941 | |
CCCCCCCCCCCCCCCCCCCCCC(=O)O |
Specifications
112-85-6 | |
White to Yellow | |
Authentic | |
C22H44O2 | |
02, 391 | |
behenic acid, 1-docosanoic acid, n-docosanoic acid, hydrofol acid 560, docosoic acid, hydrofol 2022-55, glycon b-70, behensaeure, docosanoicacid, docosansaeure | |
CCCCCCCCCCCCCCCCCCCCCC(=O)O | |
340.58 | |
CHEBI:28941 | |
85% | |
Crystalline Powder |
80.0°C to 82.0°C | |
306.0°C (60.0 mmHg) | |
80% min. (GC) | |
MFCD00002807 | |
15, 1022 | |
UKMSUNONTOPOIO-UHFFFAOYSA-N | |
docosanoic acid | |
8215 | |
340.58 | |
Technical | |
Docosanoic acid |
Safety and Handling
missing translation for 'einecsNumber' : 204-010-8
RUO – Research Use Only