missing translation for 'onlineSavingsMsg'
Learn More

Dodecacarbonyltetracobalt, 98% min, Thermo Scientific Chemicals

Product Code. 11332138
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17786-31-1
Molecular Formula C12Co4O12
Molecular Weight (g/mol) 571.853
MDL Number MFCD00064698
InChI Key YMFAWOSEDSLYSZ-UHFFFAOYSA-N
Synonym dodecacarbonyltetracobalt, dodecacarbonyltetracobalt min
PubChem CID 6096999
IUPAC Name carbon monoxide;cobalt
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Co].[Co].[Co].[Co]

Specifications

Melting Point 60°C (dec.)
Color Black
Physical Form Crystalline
Quantity 25 g
Linear Formula Co4(CO)12
UN Number UN3179
Formula Weight 571.86
Percent Purity ≤98%
Sensitivity Air Sensitive
Density 2.09g/mL
Chemical Name or Material Dodecacarbonyltetracobalt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'