missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecacarbonyltriiron, 96% (dry wt.), stab. with 5-10% methanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Description
It eliminates sulfur from substituted thiophene complexes, which leads to ferroles. Access to new areas of organometallic chemistry is promising using the reaction of Fe3(CO)12 with tellurium-nitrogen heterocycles such as benzoisotellurazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 140°C (decomposition) |
Color | Black |
Density | 2.00 g/mL |
Linear Formula | Fe3(CO)12 |
Quantity | 2 g |
UN Number | UN3179 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Insoluble in water. Soluble in non polar organic solvents. |
Formula Weight | 503.67 |
Percent Purity | 96% |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Dodecacarbonyltriiron, stab. with 5-10% methanol
- Danger
- Flammable solid Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 2
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H371-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Dodecacarbonyltriiron, Methanol
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'