missing translation for 'onlineSavingsMsg'
Learn More

Dodecanal, 95%, stab., Thermo Scientific Chemicals

Store cold

£45.53 - £126.63

Chemical Identifiers

CAS 112-54-9
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.32
MDL Number MFCD00007017
InChI Key HFJRKMMYBMWEAD-UHFFFAOYSA-N
Synonym lauraldehyde, dodecyl aldehyde, lauric aldehyde, lauryl aldehyde, n-dodecanal, dodecanaldehyde, 1-dodecanal, n-dodecyl aldehyde, n-lauraldehyde, n-dodecylic aldehydeShow MoreShow Less
PubChem CID 8194
ChEBI CHEBI:27836
IUPAC Name dodecanal
SMILES CCCCCCCCCCCC=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11437487
SDS View Documents
Thermo Scientific Alfa Aesar
A15611.22
100 g
£45.53
100g
Please to purchase this item. Need a web account? Register with us today!
11447487
SDS View Documents
Thermo Scientific Alfa Aesar
A15611.36
500 g
£126.63
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dodecyl aldehyde is used as a fragrance agent. It is used as a common ingredient in perfumery. It's end applications include soap, detergent, beauty care product, household product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecyl aldehyde is used as a fragrance agent. It is used as a common ingredient in perfumery. It′s end applications include soap, detergent, beauty care product, household product.

Solubility
Soluble in chloroform, methanol, alcohol, and propylene glycol. Insoluble in glycerol and water.

Notes
Air sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c

H315-H317-H319

missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III

missing translation for 'einecsNumber' : 203-983-6

missing translation for 'rtecsNumber' : JR9100000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dodecanal, 95%, stab., Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.