Learn More
Dodecane, mixture of isomers, Thermo Scientific Chemicals
Dodecane, C12H26, CAS Number-13475-82-6, heptane,2,2,4,6,6-pentamethyl, 2,2,4,6,6-pentamethyl-heptane, heptane, 2,2,4,6,6-pentamethyl, acmc-1c04u, vkpskydesgttfr-uhfffaoysa, permethyl 99a, 1L, 170.0 deg.C to 195.0 deg.C, CHEBI:77509, Undesignated, 0.7510g/mL, 236-757-, 44 deg.C, 170.34
£29.75 - £327.63
Chemical Identifiers
CAS | 13475-82-6 |
---|---|
Molecular Formula | C12H26 |
Molecular Weight (g/mol) | 170.34 |
MDL Number | MFCD00008969 |
InChI Key | VKPSKYDESGTTFR-UHFFFAOYSA-N |
PubChem CID | 26058 |
ChEBI | CHEBI:77509 |
IUPAC Name | 2,2,4,6,6-pentamethylheptane |
SMILES | CC(CC(C)(C)C)CC(C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10447592
|
Thermo Scientific Acros
365770010 |
1 L | Glass bottle |
£29.75
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10703413
|
Thermo Scientific Acros
365770050 |
5 L | Plastic drum |
£87.54
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10669863
|
Thermo Scientific Acros
365770250 |
25 L | Metal drum |
£327.63
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13475-82-6 | |
170.34 | |
VKPSKYDESGTTFR-UHFFFAOYSA-N | |
CHEBI:77509 | |
CC(CC(C)(C)C)CC(C)(C)C |
C12H26 | |
MFCD00008969 | |
26058 | |
2,2,4,6,6-pentamethylheptane |
Specifications
13475-82-6 | |
0.7510g/mL | |
44°C | |
C12H26 | |
MFCD00008969 | |
heptane, 2,2,4,6,6-pentamethyl, heptane,2,2,4,6,6-pentamethyl, permethyl 99a, acmc-1c04u, vkpskydesgttfr-uhfffaoysa, 2,2,4,6,6-pentamethyl-heptane | |
VKPSKYDESGTTFR-UHFFFAOYSA-N | |
2,2,4,6,6-pentamethylheptane | |
26058 | |
170.34 | |
Dodecane |
-60.0°C | |
170.0°C to 195.0°C | |
Authentic | |
1.4190 to 1.4230 | |
0.751 | |
Solubility in water: not miscible. | |
CC(CC(C)(C)C)CC(C)(C)C | |
170.34 | |
CHEBI:77509 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 236-757-
- Dodecane, mixture of isomers
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Flammable liquid Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H226-Flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
RUO – Research Use Only