missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecanedioic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
Description
Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.
Solubility
Soluble in water, ethanol, hot methanol, hot toluene and hot acetic acid.
Notes
Incompatible with strong oxidizing agents and reducing agents.
Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.
Solubility
Soluble in water, ethanol, hot methanol, hot toluene and hot acetic acid.
Notes
Incompatible with strong oxidizing agents and reducing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 127°C to 131°C |
Density | 1.15 |
Boiling Point | 244°C to 245°C (10 mmHg) |
Flash Point | 227°C (440°F) |
Odor | Odorless |
Quantity | 250 g |
Beilstein | 1782580 |
Solubility Information | Soluble in water,ethanol,hot methanol,hot toluene and hot acetic acid. |
Formula Weight | 230.31 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'