missing translation for 'onlineSavingsMsg'
Learn More
Dodecyl Sulfate, Sodium Salt 85%, Thermo Scientific Chemicals Product Code.: 10761691

Dodecyl Sulfate, Sodium Salt 85%, Thermo Scientific Chemicals

Product Code. 10761691
100 g, Plastic Bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10761691

Brand: Thermo Scientific Acros 226141000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3, 7757-82-6, 7647-14-5
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Dodecyl Sodium Sulfate, SDS, Sodium lauryl sulfate
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color White to Yellow
Physical Form Powder or Crystals
Melting Point 206.0°C
Quantity 100 g
Flash Point >150°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Assay Percent Range 85%
Linear Formula CH3(CH2)11OSO3Na
Solubility Information Solubility in water: 130g/L (20°C).
Formula Weight 288.38
Percent Purity 85%
Grade Pure
Chemical Name or Material Dodecyl sulfate, sodium salt
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457678 Certificate TypeCertificate of Analysis Date15/11/2023 Catalog Number226141000,226140025,226145000
Lot NumberA0394565 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number226141000,226140025,226145000
Lot NumberA0394564 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number226141000,226140025,226145000
Lot NumberA0414324 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number226141000,226140025,226145000
Lot NumberA0448612 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number226141000,226140025,226145000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dodecyl sulfate sodium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dodecyl Sulfate, Sodium Salt 85%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.