missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecyl Sulfate, Sodium Salt 85%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Plastic drum
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
Color | White to Yellow |
Physical Form | Powder or Crystals |
Melting Point | 206.0°C |
Quantity | 2.5 kg |
Flash Point | >150°C |
Infrared Spectrum | Authentic |
Packaging | Plastic drum |
Assay Percent Range | 85% |
Linear Formula | CH3(CH2)11OSO3Na |
Solubility Information | Solubility in water: 130g/L (20°C). |
Mostrar más |
Seguridad y manipulación
Nombre del producto
- Dodecyl sulfate sodium salt
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'