missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dodecyl sulfate, sodium salt, 99%, for biochemistry

£21.51 - £756.77

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10224183
View Documents
Thermo Scientific Chemicals
230420100
10 g
£21.51
10g
Please to purchase this item. Need a web account? Register with us today!
10569823
View Documents
Thermo Scientific Chemicals
230420250
25 g
£27.74
25g
Please to purchase this item. Need a web account? Register with us today!
10368700
View Documents
Thermo Scientific Chemicals
230421000
100 g
£62.21
100g
Please to purchase this item. Need a web account? Register with us today!
10184950
View Documents
Thermo Scientific Chemicals
230425000
500 g
£201.00
500g
Please to purchase this item. Need a web account? Register with us today!
10408093
View Documents
Thermo Scientific Chemicals
230420025
2.5 kg
£756.77
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry
Specifications

Chemical Identifiers

151-21-3
C12H25NaO4S

Specifications

151-21-3
288.38
MFCD00036175
Plastic bottle
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
Solubility in water: 130g/L (20°C). Other solubilities: partly soluble in alcohol, soluble 75g/L in ethanol (20°C)
Crystalline Powder or Crystals
Dodecyl sulfate, sodium salt
White to Beige
C12H25NaO4S
206.0°C
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
99%, Biochemical
Sodium lauryl sulfate
99%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Dodecyl sulfate sodium salt

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only