Learn More
Thermo Scientific Chemicals Dodecyl sulfate sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free
Brand: Thermo Scientific Chemicals 327310025
978.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
(0.1 M in water), (0.1 M in water) at 0nm, 0.05 max. at 260nm, 0.05 max. at 280nm | |
White | |
Powder | |
2.5 kg | |
Authentic | |
C12H25NaO4S | |
MFCD00036175 | |
Solubility in water: 130g/L (20°C). Other solubilities: partly soluble in alcohol, soluble 75g/L in ethanol (20°C) | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
CHEBI:8984 | |
≥99% |
Dodecyl sulfate, sodium salt | |
151-21-3, 7757-82-6, 7647-14-5 | |
206.0°C | |
>150°C | |
99+% | |
CH3(CH2)11OSO3Na | |
Dodecyl Sodium Sulfate, SDS, Sodium lauryl sulfate | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium dodecyl sulfate | |
3423265 | |
288.38 | |
Molecular Biology |
Safety and Handling
- Dodecyl sulfate sodium salt
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0441451 | Certificate of Analysis | 12/09/2023 | |
A0395583 | Certificate of Analysis | 14/12/2022 | |
A0391484 | Certificate of Analysis | 14/12/2022 | 327311000,327315000,327310025 |
A0402282 | Certificate of Analysis | 13/12/2022 | |
A0413136 | Certificate of Analysis | 18/04/2022 | 327311000,327315000,327310025 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only