missing translation for 'onlineSavingsMsg'
Learn More
Dodecyl sulfate, sodium salt, pure, specified according to the requirements of Ph.Eu, Thermo Scientific Chemicals Product Code.: 15509473

Dodecyl sulfate, sodium salt, pure, specified according to the requirements of Ph.Eu, Thermo Scientific Chemicals

Product Code. 15509473
1 kg
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 15509473

Brand: Thermo Scientific Acros 459000010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponalShow MoreShow Less
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium;dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Melting Point 206.0°C
Flash Point >150°C
Linear Formula CH3(CH2)11OSO3Na
Quantity 1 kg
Solubility Information Solubility in water: 130g/L (20°C).
Formula Weight 288.38
Grade Pure
Chemical Name or Material Dodecyl sulfate, sodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468888 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number459000050,459000010
Lot NumberA0467230 Certificate TypeCertificate of Analysis Date31/10/2024 Catalog Number459000050,459000010
Lot NumberA0464395 Certificate TypeCertificate of Analysis Date10/07/2024 Catalog Number459000050,459000010
Lot NumberA0458258 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number459000050,459000010
Lot NumberA0453627 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number459000050,459000010
compliance-icons compliance-icons
Product Identifier
  • Dodecyl sulfate sodium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dodecyl sulfate, sodium salt, pure, specified according to the requirements of Ph.Eu, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.