missing translation for 'onlineSavingsMsg'
Learn More

Dodecyltrimethylammonium bromide, 99%, pure, Thermo Scientific Chemicals

£60.00 - £498.48

Chemical Identifiers

CAS 1119-94-4
Molecular Formula C15H34BrN
Molecular Weight (g/mol) 308.34
InChI Key XJWSAJYUBXQQDR-UHFFFAOYSA-M
Synonym dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide
PubChem CID 14249
ChEBI CHEBI:282662
IUPAC Name dodecyl(trimethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Br-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10493043
View Documents Promotion Details
Thermo Scientific Acros
409310250
25 g
£60.00
25g
Please to purchase this item. Need a web account? Register with us today!
10687021
View Documents
Thermo Scientific Acros
409311000
100 g
£152.76
100g
Please to purchase this item. Need a web account? Register with us today!
11945451
View Documents
Thermo Scientific Acros
409315000
500 g
£498.48
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Chemical Identifiers

1119-94-4
308.34
dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide
CHEBI:282662
CCCCCCCCCCCC[N+](C)(C)C.[Br-]
C15H34BrN
XJWSAJYUBXQQDR-UHFFFAOYSA-M
14249
dodecyl(trimethyl)azanium;bromide

Specifications

1119-94-4
White to Yellow
98.5% min. (Argentometry)
C15H34BrN
dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide
XJWSAJYUBXQQDR-UHFFFAOYSA-M
dodecyl(trimethyl)azanium;bromide
14249
308.34
Pure
Dodecyltrimethylammonium bromide
246.0°C
Authentic
Glass bottle
CH3(CH2)11N(Br)(CH3)3
(10% in water) Clear to slightly hazy
CCCCCCCCCCCC[N+](C)(C)C.[Br-]
308.34
CHEBI:282662
99%
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Very toxic to aquatic life.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 214-290-3

Product Identifier
  • Dodecyltrimethylammonium bromide

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only