missing translation for 'onlineSavingsMsg'
Learn More

Dofetilide

KV11.1 (hERG) channel blocker; inhibits rapid delayed rectifier K+ current (IKr)

£114.00 - £357.00

Specifications

Chemical Name or Material 1-(4-Methanesulphonamidophenoxy)-2-[N-(4-methanesulphonamidophenethyl)-N-methylamino]ethane
CAS 115256-11-6
Assay Percent Range >99%
Molecular Formula C19H27N3O5S2
Formula Weight Observed MW: 441.56
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18737696
View Documents
Tocris Bioscience
3757/25
25 mg
£357.00
25mg
Please to purchase this item. Need a web account? Register with us today!
18716688
View Documents Promotion Details
Tocris Bioscience
3757/5
5 mg
£114.00
5mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Selective potassium channel blocker. Blocks KV11.1 (hERG) channels; inhibits the rapid delayed-rectifier K+ current (IKr). Displays class III antiarrhythmic properties.
Specifications

Specifications

1-(4-Methanesulphonamidophenoxy)-2-[N-(4-methanesulphonamidophenethyl)-N-methylamino]ethane
>99%
Observed MW: 441.56
115256-11-6
C19H27N3O5S2
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature

SDS
Documents

Documents

Product Certifications