missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Dog (Beagle) Cryopreserved Hepatocytes, Male

Product Code. 10364640
Click to view available options
Quantity:
1.5 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10364640

Brand: Gibco™ DGCS10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Dog (beagle) cryopreserved hepatocytes, male

These cryopreserved male dog (Beagle) hepatocytes have been characterized for general Phase I and Phase II drug metabolizing enzyme activities.

Cells per vial: 4-8 million

Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes, with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.

Each lot is tested for:
  • Phase I and II enzyme activities
  • Proper morphology
  • Viabilities generally >75%

    For research use only. Not intended for human or animal therapeutic or diagnostic use.

    Related Links:

    Learn more about our cryopreserved hepatocytes

    Learn about our full line of hepatocytes ,

    Note:
    These products require shipment in LN2 vapor dewars therefore additional fees may apply.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Vials of cryopreserved hepatocytes should be stored in a cryostorage dewar or -135°C freezer.
    Cell Type Hepatocytes
    Form Cryopreserved
    Gender Male
    Species Canine
    Donor Source Single Donor
    No. of Cells 4-8 x 106
    Product Line Gibco
    Product Type Hepatocytes
    Quantity 1.5 mL
    Tested For Phase I and II Enzyme Activity, Proper Morphology, Viability Generally >75%
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.