Learn More
Thermo Scientific™ Domed and Flat PCR Cap Strips
Optimize PCR applications with these cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.
£31.62 - £54.50
Product Code | Brand | Color | Format | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Format | Quantity | Price | Quantity & Availability | |||
10690174
|
Thermo Scientific™
AB0783 |
Clear | Strips of 8 Flat Caps | 120/Cs. |
£31.62
Pack of 120 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||
10249733
|
Thermo Scientific™
AB0784 |
Clear | Strips of 8 Flat Caps | 250/Cs. |
£54.50
Pack of 250 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
Description
These PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Specifications
Strips of 8 Flat Caps |