Learn More
Thermo Scientific™ Domed PCR Caps, strips of 12
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
£44.00 - £84.25
Specifications
Color Clear
Material Polypropylene
No. of Wells 12
Format Strips of 12 Domed Caps
Product Type PCR Plate Cap Strip
View More Specs
Products 2
Product Code Brand Color Format Quantity Price Quantity & Availability  
Product Code Brand Color Format Quantity Price Quantity & Availability  
11584774
View Documents Promotion Details
Thermo Scientific™
AB0852
Clear Strips of 12 Domed Caps 10 packs of 8
£44.00
Pack of 80
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
11594774
View Documents
Thermo Scientific™
AB0853
Clear Strips of 12 Domed Caps 25 packs of 8
£84.25
Pack of 200
In Stock
to see stock.
Add to basket
Description
Description
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
Specifications
Specifications
Clear
12
PCR Plate Cap Strip
96-well Plate
Polypropylene
Strips of 12 Domed Caps
Yes