Learn More
Thermo Scientific™ Domed PCR Caps, strips of 8
Click to view available options
Quantity:
10 packs of 12
250 strips
Color:
Clear
Förpackningsstorlek:
Pack of 120
Pack of 250
Beskrivning
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
Specifikationer
Specifikationer
| Color | Clear |
| Material | Polypropylene |
| No. of Wells | 8 |
| Quantity | 250 strips |
| Format | Strips of 8 Domed Caps |
| Product Type | PCR Plate Cap Strip |
| Certified RNase/DNase-free | Yes |
| Plate Compatibility | 96-well Plate |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering