missing translation for 'onlineSavingsMsg'
Learn More
Domiphen bromide, 97%, Thermo Scientific Chemicals Product Code.: 11327806

Domiphen bromide, 97%, Thermo Scientific Chemicals

Product Code. 11327806
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11327806

Brand: Thermo Scientific Alfa Aesar L12930.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.

Solubility
Soluble in ethanol, acetone, ethyl acetate, chloroform and water. Slightly soluble in benzene.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-71-6
Molecular Formula C22H40BrNO
Molecular Weight (g/mol) 414.472
MDL Number MFCD00011769
InChI Key OJIYIVCMRYCWSE-UHFFFAOYSA-M
Synonym domiphen bromide, bradasol, bradonit, bradoral, fungitex, oradol, phenododecinium bromide, bradosol bromide, fungitex r
PubChem CID 10866
IUPAC Name dodecyl-dimethyl-(2-phenoxyethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1.[Br-]

Specifications

Melting Point 117°C to 118°C
Quantity 100 g
Beilstein 3922742
Sensitivity Hygroscopic
Merck Index 14,3415
Solubility Information Soluble in ethanol,acetone,ethyl acetate,chloroform and water. Slightly soluble in benzene.
Formula Weight 414.48
Percent Purity 97%
Chemical Name or Material Domiphen bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY10I039 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog NumberL12930.14,L12930.22
Lot NumberY08H017 Certificate TypeCertificate of Analysis Date19/08/2022 Catalog NumberL12930.14,L12930.22
Lot NumberP24I010 Certificate TypeCertificate of Analysis Date14/04/2022 Catalog NumberL12930.14,L12930.22
Lot NumberE20X027 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL12930.14,L12930.22
Lot NumberW11E019 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL12930.14,L12930.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Domiphen bromide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.