missing translation for 'onlineSavingsMsg'
Learn More

Donepezil Hydrochloride, MedChemExpress

Product Code. p-200096556
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16454217

Brand: Medchem Express HYB0034

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Donepezil Hydrochloride (E2020) is a reversible, selective AChE inhibitor with an IC50 of 6.7 nM for AChE activity. Donepezil shows high selectivity for AChE over BuChE. Donepezil exhibits neuroprotective effect on Aβ42 neurotoxicity.

Donepezil Hydrochloride (E2020) is a reversible, selective AChE inhibitor with an IC50 of 6.7 nM for AChE activity. Donepezil shows high selectivity for AChE over BuChE. Donepezil exhibits neuroprotective effect on Aβ42 neurotoxicity.

Specifications

Chemical Name or Material Donepezil Hydrochloride
CAS 120011-70-3
Color White
Purity Grade Notes Research
Molecular Formula C24H30ClNO3
Quantity 50 mg
Synonym E2020
Solubility Information H2O : 33.33 mg/mL (80.13 mM; Need ultrasonic) ∣DMSO : 6.2 mg/mL (14.91 mM; Need warming)
SMILES O=C1C(CC2CCN(CC3=CC=CC=C3)CC2)CC4=C1C=C(OC)C(OC)=C4.[H]Cl
Molecular Weight (g/mol) 415.95
Formula Weight 415.95
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.