Learn More
Donkey anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, HRP, Invitrogen™
Donkey Polyclonal Secondary Antibody
Brand: Invitrogen SA1300
Code : LE
Additional Details : Weight : 0.01000kg
Description
Product SA1-300 is conjugated to HRP and optimized for ECL applications. Product SA1-300 reacts with the heavy and light chains of the immunoglobulin molecule. Pre-absorbed with the following species proteins: Bovine, goat, guinea pig, syrian hamster, horse, human, mouse, rabbit, rat, and sheep serum proteins.
Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Chicken IgY (H+L) Cross-Adsorbed | |
Polyclonal | |
HRP | |
Donkey | |
250 μL | |
Secondary | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Western Blot | |
32 μg/mL | |
PBS with 50% glycerol, proprietary stabilizer and 0.07% Kathon | |
Antigen affinity chromatography | |
RUO | |
Chicken | |
Liquid |