Learn More
Donkey anti-Chicken IgY (H+L) Secondary Antibody, FITC, Invitrogen™
Donkey Polyclonal Secondary Antibody
Brand: Invitrogen SA172000
Code : LD
Additional Details : Weight : 0.01000kg
Description
SA1-72000 detects Chicken IgY in chicken samples. SA1-72000 has been successfully used in Western Blot, and ELISA procedures. The SA1-72000 immunogen is from purified chicken IgY isolated from normal Chicken eggs.
Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Chicken IgY (H+L) | |
Polyclonal | |
FITC | |
Donkey | |
purified | |
RUO | |
Chicken | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.075% sodium azide; pH 7.2 | |
Purified chicken IgY isolated from normal chicken eggs. | |
500 μg | |
Secondary | |
-20° C, Avoid Freeze/Thaw Cycles, store in dark | |
IgG |
Safety and Handling
- Donkey anti-Chicken IgY (H+L) Secondary Antibody, FITC
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : glycerol