missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Donkey anti-Mouse IgG (H+L) ReadyProbes™ Secondary Antibody, Alexa Fluor™ 594

Donkey Polyclonal Secondary Antibody

Brand:  Invitrogen™ R37115

174.80 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 15126896

  • £188.00 / 2 ampoules

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

No pipetting or dilutions required. Just tip and drip two drops per mL buffer to stain your cells. This antibody shows minimum cross-reactivity to bovine, chicken, goat, guinea pig, hamster, horse, human, rabbit, rat, and sheep serum proteins. Like other ReadyProbes reagents, Alexa Fluor 594 Donkey anti-Mouse IgG Antibody comes in a convenient dropper bottle.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse IgG (H+L)
Polyclonal
PBS with 0.1% BSA and <0.1% sodium azide
Gamma Immunoglobin.
2 x 2.5 mL
Secondary
4°C, store in dark
Liquid
Western Blot, Immunocytochemistry, Flow Cytometry
Alexa Fluor 594
Donkey
Affinity chromatography
RUO
Mouse
Secondary Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Donkey anti-Mouse IgG (H+L) ReadyProbes™ Secondary Antibody, Alexa Fluor™ 594 > 2 x 2.5 mL; Alexa Fluor 594

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.